Learn More
VAMP5, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant VAMP5.
Brand: Abnova H00010791-A01.50uL
Description
Synaptobrevins/VAMPs, syntaxins, and the 25kDa synaptosomal-associated protein are the main components of a protein complex involved in the docking and/or fusion of vesicles and cell membranes. The VAMP5 gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family and the SNARE superfamily. This VAMP family member may participate in vesicle trafficking events that are associated with myogenesis. [provided by RefSeq
Sequence: MAGIELERCQQQANEVTEIMRNNFGKVLERGVKLAELQQRSDQLLDMSSTFNKTTQNLAQKKCWENIRYRSpecifications
| VAMP5 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant VAMP5. | |
| VAMP5 | |
| VAMP5 | |
| VAMP5 (NP_006625, 1 a.a. ∽ 70 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 10791 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_006634 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only