Learn More
UNG, Mouse, Clone: k1C12, Abnova™
Mouse monoclonal antibody raised against full-length recombinant UNG.
Brand: Abnova MAB1099.100uL
Description
This gene encodes one of several uracil-DNA glycosylases. One important function of uracil-DNA glycosylases is to prevent mutagenesis by eliminating uracil from DNA molecules by cleaving the N-glycosylic bond and initiating the base-excision repair (BER) pathway. Uracil bases occur from cytosine deamination or misincorporation of dUMP residues. Alternative promoter usage and splicing of this gene leads to two different isoforms: the mitochondrial UNG1 and the nuclear UNG2. [provided by RefSeq
Specifications
| UNG | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user. | |
| UNG | |
| DGU/DKFZp781L1143/HIGM4/UDG/UNG1/UNG15/UNG2 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| k1C12 | |
| Mouse monoclonal antibody raised against full length recombinant UNG. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| NM_080911 | |
| UNG | |
| Recombinant protein corresponding to full length human UNG. | |
| RUO | |
| 7374 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.