Learn More
TRIM16, Mouse, Clone: 5G11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant TRIM16.
Brand: Abnova H00010626-M02A.200uL
Description
This gene was identified as an estrogen and anti-estrogen regulated gene in epithelial cells stably expressing estrogen receptor. The protein encoded by this gene contains two B box domains and a coiled-coiled region that are characteristic of the B box zinc finger protein family. The proteins of this family have been reported to be involved in a variety of biological processes including cell growth, differentiation and pathogenesis. Expression of this gene was detected in most tissues. Its function, however, has not yet been determined. [provided by RefSeq
Sequence: LDAARRDKEAELQCTQLDLERKLKLNENAISRLQANQKSVLVSVSEVKAVAEMQFGELLAAVRKAQANVMLFLEEKEQAALSQANGIKAHLEYRSAEMEKSKQELERMASpecifications
| TRIM16 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_006470 | |
| TRIM16 | |
| TRIM16 (NP_006461,165 a.a. ∽ 273 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10626 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 5G11 | |
| Mouse monoclonal antibody raised against a partial recombinant TRIM16. | |
| TRIM16 | |
| EBBP | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.