missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOPORS, Mouse, Clone: 6D2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant TOPORS.
Brand: Abnova H00010210-M15A.200uL
This item is not returnable.
View return policy
Description
Sequence: SPDSKCPICLDRFDNVSYLDRCLHKFCFRCVQEWSKNKAECPLCKQPFDSIFHSVRAEDDFKEYVLRPSYNGSFVTPDRRFRYRTTLTRERNASVYSPSGPVNRRTTTSpecifications
| TOPORS | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_005802 | |
| TOPORS | |
| TOPORS (NP_005793,98 a.a. ∽ 205 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10210 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 6D2 | |
| Mouse monoclonal antibody raised against a partial recombinant TOPORS. | |
| TOPORS | |
| LUN/P53BP3/RP31/TP53BPL | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction