Learn More
TIMP1, Mouse, Clone: 2A5, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of TIMP1.
Brand: Abnova MAB1799.200ug
Description
This gene belongs to the TIMP gene family. The proteins encoded by this gene family are natural inhibitors of the matrix metalloproteinases (MMPs), a group of peptidases involved in degradation of the extracellular matrix. In addition to its inhibitory role against most of the known MMPs, the encoded protein is able to promote cell proliferation in a wide range of cell types, and may also have an anti-apoptotic function. Transcription of this gene is highly inducible in response to many cytokines and hormones. In addition, the expression from some but not all inactive X chromosomes suggests that this gene inactivation is polymorphic in human females. This gene is located within intron 6 of the synapsin I gene and is transcribed in the opposite direction. [provided by RefSeq
Specifications
| TIMP1 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| TIMP1 | |
| TIMP1 | |
| A synthetic peptide corresponding to human TIMP1. | |
| RUO | |
| 7076 | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 2A5 | |
| Mouse monoclonal antibody raised against synthetic peptide of TIMP1. | |
| In PBS (0.08% sodium azide) | |
| CLGI/EPA/EPO/FLJ90373/HCI/TIMP | |
| Mouse | |
| 200 μg | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.