Learn More
TGIF2, Mouse, Clone: 3G8, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant TGIF2.
Brand: Abnova H00060436-M13A.200uL
Description
The protein encoded by this gene is a DNA-binding homeobox protein and a transcriptional repressor. The encoded protein appears to repress transcription by recruiting histone deacetylases to TGF beta-responsive genes. This gene is amplified and overexpressed in some ovarian cancers, and mutations in this gene can cause holoprosencephaly. [provided by RefSeq
Sequence: MSDSDLGEDEGLLSLAGKRKRRGNLPKESVKILRDWLYLHRYNAYPSEQEKLSLSGQTNLSVLQICNWFINARRRLLPDMLRKDGKDPNQFTISRRGGKASDVALPRGSSPSVLAVSVPAPTNVLSLSVCSMPLHSGQGEKPAAPFPRGELESPKPLVTPGSTLTLLTRAEAGSPTGGLFNTPPPTPPEQDKEDFSSFQLLVEVALQRAAEMELQKQQDPSLPLLHTPIPLVSENPQSpecifications
| TGIF2 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC012816 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Western Blot | |
| 3G8 | |
| Mouse monoclonal antibody raised against a full-length recombinant TGIF2. | |
| TGIF2 | |
| TGIF2 | |
| TGIF2 (AAH12816,1 a.a. ∽ 237 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 60436 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.