Learn More
TF, Mouse, Clone: 11D3, Abnova™
Mouse monoclonal antibody raised against native TF.
Brand: Abnova MAB3056.200ug
Description
This gene encodes a glycoprotein with an approximate molecular weight of 76.5kDa. It is thought to have been created as a result of an ancient gene duplication event that led to generation of homologous C and N-terminal domains each of which binds one ion of ferric iron. The function of this protein is to transport iron from the intestine, reticuloendothelial system, and liver parenchymal cells to all proliferating cells in the body. This protein may also have a physiologic role as granulocyte/pollen-binding protein (GPBP) involved in the removal of certain organic matter and allergens from serum. [provided by RefSeq
Specifications
| TF | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| TF | |
| TF | |
| Native purified transferrin from pooled human plasma. | |
| RUO | |
| 7018 | |
| Store at 4°C. | |
| IgG1 |
| ELISA | |
| 11D3 | |
| Mouse monoclonal antibody raised against native TF. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| DKFZp781D0156/PRO1557/PRO2086 | |
| Mouse | |
| 200 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.