Learn More
TECTA, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant TECTA.
Brand: Abnova H00007007-A01.50uL
Description
The tectorial membrane is an extracellular matrix of the inner ear that contacts the stereocilia bundles of specialized sensory hair cells. Sound induces movement of these hair cells relative to the tectorial membrane, deflects the stereocilia, and leads to fluctuations in hair-cell membrane potential, transducing sound into electrical signals. Alpha-tectorin is one of the major noncollagenous components of the tectorial membrane. Mutations in the TECTA gene have been shown to be responsible for autosomal dominant nonsyndromic hearing impairment and a recessive form of sensorineural pre-lingual nonsyndromic deafness. [provided by RefSeq
Sequence: YATPTRDSNDKLRYFIIEGGCQNLKDNTIGIEENAVSLTCRFHVTVFKFIGDYDEVHLHCAVSLCDSEKYSCKITCPHNSRIATDYTKEPKEQIISVGPISpecifications
| TECTA | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant TECTA. | |
| TECTA | |
| DFNA12/DFNA8/DFNB21 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_005422 | |
| TECTA | |
| TECTA (NP_005413, 1981 a.a. ∽ 2080 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 7007 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only