Learn More
TEAD3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant TEAD3.
Brand: Abnova H00007005-A01.50uL
Description
This gene product is a member of the transcriptional enhancer factor (TEF) family of transcription factors, which contain the TEA/ATTS DNA-binding domain. It is predominantly expressed in the placenta and is involved in the transactivation of the chorionic somatomammotropin-B gene enhancer. Translation of this protein is initiated at a non-AUG (AUA) start codon. [provided by RefSeq
Sequence: WQDRTIASSRLRLLEYSAFMEVQRDPDTYSKHLFVHIGQTNPAFSDPPLEAVDVRQIYDKFPEKKGGLKELYEKGPPNAFFLVKFWADSpecifications
| TEAD3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant TEAD3. | |
| TEAD3 | |
| DTEF-1/ETFR-1/TEAD5/TEF-5/TEF5 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_003214 | |
| TEAD3 | |
| TEAD3 (NP_003205, 215 a.a. ∽ 302 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 7005 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only