Learn More
TCN1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant TCN1.
Brand: Abnova H00006947-A01.50uL
Description
This gene encodes a member of the vitamin B12-binding protein family. This family of proteins, alternatively referred to as R binders, is expressed in various tissues and secretions. This protein is a major constituent of secondary granules in neutrophils and facilitates the transport of cobalamin into cells. [provided by RefSeq
Sequence: MRQSHQLPLVGLLLFSFIPSQLCEICEVSEENYIRLKPLLNTMIQSNYNRGTSAVNVVLSLKLVGIQIQTLMQKMIQQIKYNVKSRLSDVSSGELALIILALGVCRNAEENLIYDYHLIDKLENKFQAEIENMEAHNGTPLTNYYQLSLDVLALCLFNGNYSTAEVVNHFTPENKNYYFGSQFSVDTGAMAVLALTCVKKSLINGQIKADEGSLKNISIYTKSLVEKILSEKKENGLIGNTFSTGEAMQALFVSSDYYNENDWNCRQTLNTVLTEISQGAFSNPNAAAQVLPALMGKTFLDINKDSSCVPASGNFNISADEPITVTPPDSQSYISVDYSVRINETYFTNVAVLNGSVFLSVMEKAQKMNDTIFGFTMEERSWGPYITCIQGLCANNNDRTYWELLSGGEPLSQGAGSYVVRNGSpecifications
| TCN1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length recombinant TCN1. | |
| TCN1 | |
| TC1/TCI | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA | |
| Unconjugated | |
| 50% glycerol | |
| BC018632 | |
| TCN1 | |
| TCN1 (AAH18632, 1 a.a. ∽ 423 a.a) full-length recombinant protein with GST tag. | |
| RUO | |
| 6947 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only