missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Streptavidin, Mouse, Clone: 1C2, Abnova™
Mouse monoclonal antibody raised against partial recombinant Streptomyces avidinii Streptavidin.
Brand: Abnova MAB2046.100uL
This item is not returnable.
View return policy
Specifications
| Streptavidin | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μL | |
| Primary | |
| Liquid |
| ELISA, Western Blot | |
| 1C2 | |
| Mouse monoclonal antibody raised against partial recombinant Streptomyces avidinii Streptavidin. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| Recombinant protein corresponding to amino acids 25-183 of Streptomyces avidinii Streptavidin. | |
| RUO | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction