Learn More
STAT3, Mouse, Clone: 3B5, Abnova™
Mouse monoclonal antibody raised against recombinant STAT3.
Brand: Abnova MAB10397.100uL
Description
The protein encoded by this gene is a member of the STAT protein family. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein is activated through phosphorylation in response to various cytokines and growth factors including IFNs, EGF, IL5, IL6, HGF, LIF and BMP2. This protein mediates the expression of a variety of genes in response to cell stimuli, and thus plays a key role in many cellular processes such as cell growth and apoptosis. The small GTPase Rac1 has been shown to bind and regulate the activity of this protein. PIAS3 protein is a specific inhibitor of this protein. Three alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq
Specifications
| STAT3 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| STAT3 | |
| STAT3 | |
| Recombinant protein corresponding to human STAT3. | |
| RUO | |
| 6774 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 3B5 | |
| Mouse monoclonal antibody raised against recombinant STAT3. | |
| In ascites (0.03% sodium azide) | |
| APRF/FLJ20882/HIES/MGC16063 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Monkey, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.