missing translation for 'onlineSavingsMsg'
Learn More

STAM, Mouse, Polyclonal Antibody, Abnova™

Product Code. 16162242
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16162242 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16162242 Supplier Abnova Supplier No. H00008027A01.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a full-length recombinant STAM.

This gene was identified by the rapid tyrosine-phosphorylation of its product in response to cytokine stimulation. The encoded protein contains a SH3 domain and the immunoreceptor tyrosine-based activation motif (ITAM). This protein associates with JAK3 and JAK2 kinases via its ITAM region, and is phosphorylated by the JAK kinases upon cytokine stimulation, which suggests the function of this protein is as an adaptor molecule involved in the downstream signaling of cytokine receptors. HGS/HRS (hepatocyte grwoth factor-regulated tyrosine kinase substrate) has been found to bind and counteract the function of this protein. [provided by RefSeq

Sequence: MPLFATNPFDQDVEKATSEMNTAEDWGLILDICDKVGQSRTGPKDCLRSIMRRVNHKDPHVAMQALTLLGACVSNCGKIFHLEVCSRDFASEVSNVLNKGHPKVCEKLKALMVEWTDEFKNDPQLSLISAMIKNLKEQGVTFPAIGSQAAEQAKASPALVAKDPGTVANKKEEEDLAKAIELSLKEQRQQSTTLSTLYPSTSSLLTNHQHEGRKVRAIYDFEAAEDNELTFKAGEIITVLDDSDPNWWKGETHQGIGLFPSNFVTAYLTAEPEMIKTEKKTVQFSDDVQVETIEPEPEPAFIDEDKMDQLLQMLQSTDPSDDQPDLPELLHLEAMCHQMGPLIDEKLEDIDRKHSELSELNVKVMEALSLYTKLMNEDPMYSMYAKLQNQPGSGPTIRKPSPS

Specifications

Antigen STAM
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant STAM.
Formulation 50% glycerol
Gene STAM
Gene Accession No. BC030586
Gene Alias DKFZp686J2352/STAM1
Gene Symbols STAM
Host Species Mouse
Immunogen STAM (AAH30586, 1 a.a. ∽ 403 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8027
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.