Learn More
SRC, Mouse, Clone: 4F1E8, Abnova™
Mouse monoclonal antibody raised against partial recombinant SRC.
Brand: Abnova MAB10411.100uL
Description
This gene is highly similar to the v-src gene of Rous sarcoma virus. This proto-oncogene may play a role in the regulation of embryonic development and cell growth. The protein encoded by this gene is a tyrosine-protein kinase whose activity can be inhibited by phosphorylation by c-SRC kinase. Mutations in this gene could be involved in the malignant progression of colon cancer. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Specifications
| SRC | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| SRC | |
| SRC | |
| Recombinant protein corresponding to amino acids 10-193 of human SRC. | |
| RUO | |
| 6714 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
| ELISA, Western Blot | |
| 4F1E8 | |
| Mouse monoclonal antibody raised against partial recombinant SRC. | |
| In ascites (0.03% sodium azide) | |
| ASV/SRC1/c-SRC/p60-Src | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.