Learn More
SPN, Mouse, APC, Clone: DF-T1, Abnova™
Mouse monoclonal antibody raised against native SPN.
Brand: Abnova MAB6062.100Reactions
Description
Sialophorin (leukosialin) is a major sialoglycoprotein on the surface of human T lymphocytes, monocytes, granulocytes, and some B lymphocytes, which appears to be important for immune function and may be part of a physiologic ligand-receptor complex involved in T-cell activation.[supplied by OMIM
Specifications
| SPN | |
| Monoclonal | |
| APC | |
| Flow Cytometry (10 ul/106 cells) The optimal working dilution should be determined by the end user. | |
| SPN | |
| SPN | |
| Native purified from myeloblastic KG1 Cells. | |
| RUO | |
| 6693 | |
| Human | |
| Liquid |
| Flow Cytometry, Immunohistochemistry | |
| DF-T1 | |
| Mouse monoclonal antibody raised against native SPN. | |
| In PBS (0.09% sodium azide) | |
| CD43/GPL115/LSN | |
| Mouse | |
| 100 Reactions | |
| Primary | |
| Specificity human CD43 | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.