Learn More
SPARC, Mouse, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against native SPARC.
Brand: Abnova MAB0278.100ug
Description
Secreted protein acidic and rich in cysteine/osteonectin/BM40, or SPARC, is a matrix-associated protein that elicits changes in cell shape, inhibits cell-cycle progression, and influences the synthesis of extracellular matrix (ECM) (Bradshaw et al., 2003 [PubMed 12721366]).[supplied by OMIM
Specifications
| SPARC | |
| Monoclonal | |
| Mouse monoclonal antibody raised against native SPARC. | |
| In PBS (50% glycerol) | |
| ON | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody recognizes human, rat and bovine bone osteonectin, human platelet osteonectin. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry, Radioimmune Assays (RIA), Western Blot | |
| Unconjugated | |
| Western Blot (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| SPARC | |
| SPARC | |
| Native purified Human SPARC. | |
| RUO | |
| 6678 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.