Learn More
SPAG16, Mouse, Clone: 1D2, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant SPAG16.
Brand: Abnova H00079582-M01.100ug
Description
Cilia and flagella are comprised of a microtubular backbone, the axoneme, which is organized by the basal body and surrounded by plasma membrane. SPAG16 encodes 2 major proteins that associate with the axoneme of sperm tail and the nucleus of postmeiotic germ cells, respectively (Zhang et al., 2007 [PubMed 17699735]).[supplied by OMIM
Sequence: MAAQRGMPSSAVRVLEEALGMGLTAAGDARDTADAVAAEGAYYLEQVTITEASEDDYEYEEIPDDNFSIPEGEEDLAKAIQMAQEQATDTEILERKTVLPSKHAVPEVIEDFLCNFLIKMGMTRTLDCFQSEWYELIQKGVTELRTVGNVPDVYTQIMLLENENKNLKKDLKHYKQAAEYVIFSpecifications
| SPAG16 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001025436.1 | |
| SPAG16 | |
| SPAG16 (NP_001020607.1,1 a.a. ∽ 183 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 1D2 | |
| Mouse monoclonal antibody raised against a full-length recombinant SPAG16. | |
| SPAG16 | |
| DKFZp666P1710/FLJ22724/FLJ37717/MGC87036/PF20/WDR29 | |
| Mouse | |
| Protein A Purification | |
| RUO | |
| 79582 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.