Learn More
SORL1, Mouse, Clone: 7D7B11, Abnova™
Mouse monoclonal antibody raised against partial recombinant SORL1.
Brand: Abnova MAB10367.100uL
Description
This gene encodes a protein that belongs to the families of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins, of low density lipoprotein receptor (LDLR) proteins, and of fibronectin type III repeats proteins. In addition to VPS10, LDLR and fibronectin type 3 domains, this protein also includes an epidermal growth factor precursor-like module, a single transmembrane segment and a cytoplasmic tail with features similar to endocytosis- and sorting-competent receptors. Members of the VPS10 domain-containing receptor family are large with many exons but the CDS lengths are usually less than 3700 nt; this gene is an exception to the pattern with a CDS length greater than 6600 nt. Very large introns typically separate the exons encoding the VPS10 domain; the remaining exons are separated by much smaller-sized introns. The encoded protein is mainly intracellular and localizes in the paranuclear compartment. It is synthesized as a preproprotein, and when the propeptide is still attached, no binding occurs to the VPS10 domain. This gene is strongly expressed in the central nervous system. [provided by RefSeq
Specifications
| SORL1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| SORL1 | |
| SORL1 | |
| Recombinant protein corresponding to amino acids 2159-2214 of human SORL1. | |
| RUO | |
| 6653 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Western Blot | |
| 7D7B11 | |
| Mouse monoclonal antibody raised against partial recombinant SORL1. | |
| In ascites (0.03% sodium azide) | |
| C11orf32/FLJ21930/FLJ39258/LR11/LRP9/SORLA/SorLA-1/gp250 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.