Learn More
SOD1, Mouse, Clone: 24, Abnova™
Mouse monoclonal antibody raised against full-length recombinant SOD1.
Brand: Abnova MAB9887.100ug
Description
The protein encoded by this gene binds copper and zinc ions and is one of two isozymes responsible for destroying free superoxide radicals in the body. The encoded isozyme is a soluble cytoplasmic protein, acting as a homodimer to convert naturally-occuring but harmful superoxide radicals to molecular oxygen and hydrogen peroxide. The other isozyme is a mitochondrial protein. Mutations in this gene have been implicated as causes of familial amyotrophic lateral sclerosis. Rare transcript variants have been reported for this gene. [provided by RefSeq
Specifications
| SOD1 | |
| Monoclonal | |
| Unconjugated | |
| Western blot (1:1000) The optimal working dilution should be determined by the end user. | |
| SOD1 | |
| ALS/ALS1/IPOA/SOD/homodimer | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 6647 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 24 | |
| Mouse monoclonal antibody raised against full length recombinant SOD1. | |
| In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300) | |
| NM_000454.4 | |
| SOD1 | |
| Recombinant protein corresponding to full length human SOD1. | |
| 100 μg | |
| Primary | |
| It can expression in HeLa whole cell lysate. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.