Learn More
SNX9, Mouse, Clone: 3C6, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant SNX9.
Brand: Abnova H00051429-M03A.200uL
Description
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members, but does contain a SH3 domain near its N-terminus. This protein interacts with the cytoplasmic domains of the precursor but not the processed forms of a disintegrin and metalloprotease domain 9 and 15. This protein binds the beta-appendage domain of adaptor protein 2 and may function to assist adaptor protein 2 in its role at the plasma membrane. This protein interacts with activated Cdc42-associated kinase-2 to regulate the degradation of epidermal growth factor receptor protein. [provided by RefSeq
Sequence: MATKARVMYDFAAEPGNNELTVNEGEIITITNPDVGGGWLEGRNIKGERGLVPTDYVEILPSDGKDQFSCGNSVADQAFLDSLSASTAHASSSAASNNHQVGSGNDPWSAWSASKSGNWESSEGWGAQPEGAGAQRNTNTPNNWDTAFGHPQAYQGPATGDDDDWDEDWDGPKSSSYFKDSESADAGGAQRGNSRASSSSMKIPLNKFPGFAEPGTEQYLLAKQLAKPKEKIPIIVGDYGPMWVYPTSTFDCVVADPRKGSKMYGLKSYIEYQLTPTNTNRSVNHRYKHFDWLYERLLVKFGSAIPIPSLPDKQVTGRFEEEFIKMRMERLQAWMTRMCRHPVISESEVFQQFLNFRDEKEWKTGKRKAERDELAGVMIFSTMEPEAPDLDLVEIEQKCEAVGKFTKAMDDGVKELLTVGQEHWKRCTGPLPKEYQKIGKALQSLATVFSSSGYQGETDLNDAITEAGKTYEEIASLVAEQPKKDLHFLMECNHEYKGFLGCFPDVIGTHKGAIEKVKESDKLVATSKITLQDKQNMVKRVSIMSYALQAEMNHFHSNRIYDYNSVIRLYLEQQVQFYETIAEKLRQALSRFPVMSpecifications
| SNX9 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC005022 | |
| SNX9 | |
| SNX9 (AAH05022,1 a.a. ∽ 595 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 51429 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 3C6 | |
| Mouse monoclonal antibody raised against a full-length recombinant SNX9. | |
| SNX9 | |
| MST155/MSTP155/SDP1/SH3PX1/SH3PXD3A/WISP | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.