Learn More
SNFT, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SNFT.
Brand: Abnova H00055509-A01.50uL
Description
This gene encodes a member of the basic leucine zipper protein family. The encoded protein functions as a transcriptional repressor when heterodimerizing with JUN. The protein may play a role in repression of interleukin-2 and matrix metalloproteinase-1 transcription
Sequence: AAQRSRKKQTQKADKLHEEYESLEQENTMLRREIGKLTEELKHLTEALKEHEKMCPLLLCPMNFVPVPPRPDPVAGCLSpecifications
| SNFT | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SNFT. | |
| BATF3 | |
| JDP1/JUNDM1/SNFT | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_018664 | |
| BATF3 | |
| SNFT (NP_061134, 48 a.a. to 125 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 55509 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only