Learn More
SNCA, Mouse, Clone: 2A7, Abnova™
Mouse monoclonal antibody raised against recombinant SNCA.
Brand: Abnova MAB5383.100uL
Description
Alpha-synuclein is a member of the synuclein family, which also includes beta- and gamma-synuclein. Synucleins are abundantly expressed in the brain and alpha- and beta-synuclein inhibit phospholipase D2 selectively. SNCA may serve to integrate presynaptic signaling and membrane trafficking. Defects in SNCA have been implicated in the pathogenesis of Parkinson disease. SNCA peptides are a major component of amyloid plaques in the brains of patients with Alzheimer's disease. Four alternatively spliced transcripts encoding two different isoforms have been identified for this gene. [provided by RefSeq]
Specifications
| SNCA | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:10000) Immunofluorescence (1:1000) The optimal working dilution should be determined by the end user. | |
| SNCA | |
| SNCA | |
| Recombinant His fusion protein corresponding to human SNCA. | |
| RUO | |
| 6622 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 2A7 | |
| Mouse monoclonal antibody raised against recombinant SNCA. | |
| In ascites (5mM sodium azide) | |
| MGC110988/NACP/PARK1/PARK4/PD1 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mammalia | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.