missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMAD4, Mouse, Clone: 4G1C6, Abnova™
Mouse monoclonal antibody raised against recombinant SMAD4.
Brand: Abnova MAB10392.100uL
This item is not returnable.
View return policy
Specifications
| SMAD4 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. | |
| SMAD4 | |
| SMAD4 | |
| Recombinant protein corresponding to human SMAD4. | |
| RUO | |
| 4089 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 4G1C6 | |
| Mouse monoclonal antibody raised against recombinant SMAD4. | |
| In ascites (0.03% sodium azide) | |
| DPC4/JIP/MADH4 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction