Learn More
SMAD2, Mouse, Clone: 5G7, Abnova™
Mouse monoclonal antibody raised against recombinant SMAD2.
Brand: Abnova MAB10391.100uL
Description
The protein encoded by this gene belongs to the SMAD, a family of proteins similar to the gene products of the Drosophila gene 'mothers against decapentaplegic' (Mad) and the C. elegans gene Sma. SMAD proteins are signal transducers and transcriptional modulators that mediate multiple signaling pathways. This protein mediates the signal of the transforming growth factor (TGF)-beta, and thus regulates multiple cellular processes, such as cell proliferation, apoptosis, and differentiation. This protein is recruited to the TGF-beta receptors through its interaction with the SMAD anchor for receptor activation (SARA) protein. In response to TGF-beta signal, this protein is phosphorylated by the TGF-beta receptors. The phosphorylation induces the dissociation of this protein with SARA and the association with the family member SMAD4. The association with SMAD4 is important for the translocation of this protein into the nucleus, where it binds to target promoters and forms a transcription repressor complex with other cofactors. This protein can also be phosphorylated by activin type 1 receptor kinase, and mediates the signal from the activin. Alternatively spliced transcript variants encoding the same protein have been observed. [provided by RefSeq]
Specifications
| SMAD2 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. | |
| SMAD2 | |
| SMAD2 | |
| Recombinant protein corresponding to human SMAD2. | |
| RUO | |
| 4087 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 5G7 | |
| Mouse monoclonal antibody raised against recombinant SMAD2. | |
| In ascites (0.03% sodium azide) | |
| JV18/JV18-1/MADH2/MADR2/MGC22139/MGC34440/hMAD-2/hSMAD2 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.