Learn More
SLC22A3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SLC22A3.
Brand: Abnova H00006581-A01.50uL
Description
Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. [provided by RefSeq
Sequence: CQRYLLEAANDSASATSALSCADPLAAFPNRSAPLVPCRGGWRYAQAHSTIVSEFDLVCVNAWMLDSpecifications
| SLC22A3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SLC22A3. | |
| SLC22A3 | |
| EMT/EMTH/OCT3 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_021977 | |
| SLC22A3 | |
| SLC22A3 (NP_068812, 90 a.a. ∽ 155 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 6581 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only