missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Slc17a6, Mouse, R-PE, Clone: S29-29, Abnova™
Mouse monoclonal antibody raised against recombinant of Slc17a6.
Brand: Abnova MAB11546.100ug
This item is not returnable.
View return policy
Specifications
| Slc17a6 | |
| Monoclonal | |
| R-PE | |
| Immunohistochemistry Western blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Slc17a6 | |
| Slc17a6 | |
| Recombinant fusion protien corresponding to amino acids 501-582 at C-terminus of rat Slc17a6. | |
| 100 μg | |
| Primary | |
| Detects ∼60 kDa. | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Immunohistochemistry, Western Blot | |
| S29-29 | |
| Mouse monoclonal antibody raised against recombinant of Slc17a6. | |
| In PBS, pH 7.4 (50% glycerol) | |
| Dnpi/Vglut2 | |
| Mouse | |
| Protein G Purified | |
| RUO | |
| 84487 | |
| Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction