Learn More
SLC13A3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SLC13A3.
Brand: Abnova H00064849-A01.50uL
Description
Mammalian sodium-dicarboxylate cotransporters transport succinate and other Krebs cycle intermediates. They fall into 2 categories based on their substrate affinity: low affinity and high affinity. Both the low- and high-affinity transporters play an important role in the handling of citrate by the kidneys. The protein encoded by this gene represents the high-affinity form. Alternatively spliced transcript variants encoding different isoforms have been found for this gene, although the full-length nature of some of them have not been characterized yet. [provided by RefSeq
Sequence: LPIANAILKSLFGQKEVRKDPSQESEENTAAVRRNGLHTVPTEMQFLASTEAKDHPGETEVPLDLPADSRKEDEYRRNIWKSpecifications
| SLC13A3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SLC13A3. | |
| SLC13A3 | |
| NADC3/SDCT2 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_022829 | |
| SLC13A3 | |
| SLC13A3 (NP_073740, 152 a.a. ∽ 232 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 64849 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only