missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIT1, Mouse, PE, Clone: SIT-01, Abnova™
Mouse monoclonal antibody raised against partial recombinant SIT1.
Brand: Abnova MAB4525.100ug
This item is not returnable.
View return policy
Specifications
| SIT1 | |
| Monoclonal | |
| 0.1 mg/mL | |
| Mouse monoclonal antibody raised against partial recombinant SIT1. | |
| In PBS (0.2% BSA, 0.09% sodium azide) | |
| MGC125908/MGC125909/MGC125910/RP11-331F9.5/SIT | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody reacts with SHP2-interacting transmembrane adaptor protein (SIT) expressed exclusively in lymphoid organs. It weakly crossreacts with mouse SIT. | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Flow Cytometry, Immunoprecipitation, Western Blot | |
| SIT-01 | |
| PE | |
| Flow Cytometry (1:50) The optimal working dilution should be determined by the end user. | |
| SIT1 | |
| SIT1 | |
| Recombinant protein corresponding to partial human SIT1. | |
| RUO | |
| 27240 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction