missing translation for 'onlineSavingsMsg'
Learn More

SIPA1, Mouse, Clone: 2B4, Abnova™

Mouse monoclonal antibody raised against a partial recombinant SIPA1.

Brand:  Abnova H00006494-M01A.200uL

Product Code. 16175305

  • £303.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The product of this gene is a mitogen induced GTPase activating protein (GAP). It exhibits a specific GAP activity for Ras-related regulatory proteins Rap1 and Rap2, but not for Ran or other small GTPases. This protein may also hamper mitogen-induced cell cycle progression when abnormally or prematurely expressed. It is localized to the perinuclear region. Two alternatively spliced variants encoding the same isoform have been characterized to date. [provided by RefSeq

Sequence: SEIASTCNTILESLSREGQPIPESGDPKGTPKSDAEPEPGNLSEKVSHLESMLRKLQEDLQKEKADRAALEEEVRSLRHNNRRLQAESESAATRLLLASKQLGSPTADLA
Specifications

Specifications

SIPA1
Monoclonal
Unconjugated
ascites with no preservative
BC010492
SIPA1
SIPA1 (AAH10492,933 a.a. ∽ 1042 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
RUO
6494
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Ascites
ELISA
2B4
Mouse monoclonal antibody raised against a partial recombinant SIPA1.
SIPA1
MGC102688/MGC17037/SPA1
Mouse
200 μL
Primary
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.