missing translation for 'onlineSavingsMsg'
Learn More

SIGLEC1, Mouse, Clone: HSn 7D2, Abnova™

Product Code. 16087086
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16087086 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16087086 Supplier Abnova Supplier No. MAB7150.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against SIGLEC1.

This gene encodes a member of the immunoglobulin superfamily. The encoded protein is a lectin-like adhesion molecule that binds glycoconjugate ligands on cell surfaces in a sialic acid-dependent manner. It is a type I transmembrane protein expressed only by a subpopulation of macrophages and is involved in mediating cell-cell interactions. Alternative splicing produces a transcript variant encoding an isoform that is soluble rather than membrane-bound; however, the full-length nature of this variant has not been determined. [provided by RefSeq

Specifications

Antigen SIGLEC1
Applications ELISA, Flow Cytometry, Immunohistochemistry, Radioimmune Assays (RIA)
Classification Monoclonal
Clone HSn 7D2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against SIGLEC1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene SIGLEC1
Gene Accession No. Q9BZZ2
Gene Alias CD169/DKFZp667F058/FLJ00051/FLJ00055/FLJ00073/FLJ00411/FLJ32150/SIGLEC-1/SN/dJ1009E24.1
Gene Symbols SIGLEC1
Host Species Mouse
Immunogen Full length human SIGLEC1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6614
Test Specificity Sialoadhesin (HSn 7D2). Theoretical Molecular Weight: 183 KDa. The epitope is located within domains 1-4.
Target Species Human
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.