missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH2D3C, Mouse, Clone: 3B2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SH2D3C.
Brand: Abnova H00010044-M01A.200uL
This item is not returnable.
View return policy
Description
Sequence: MTAVGRRCPALGSRGAAGEPEAGSDYVKFSKEKYILDSSPEKLHKELEEELKLSSTDLRSHAWYHGRIPREVSETLVQRNGDFLIRDSLTSLGDYVLTCRWRNQALHFKISpecifications
| SH2D3C | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_005489 | |
| SH2D3C | |
| SH2D3C (NP_005480,1 a.a. ∽ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10044 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 3B2 | |
| Mouse monoclonal antibody raised against a partial recombinant SH2D3C. | |
| SH2D3C | |
| CHAT/FLJ39664/NSP3/PRO34088 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction