Learn More
SEMG2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human SEMG2 protein.
Brand: Abnova H00006407-B01P.50ug
Description
The secreted protein encoded by this gene is involved in the formation of a gel matrix that encases ejaculated spermatozoa. Proteolysis by the prostate-specific antigen (PSA) breaks down the gel matrix and allows the spermatozoa to move more freely. The encoded protein is found in lesser abundance than a similar semenogelin protein. The genes encoding these two semenogelin proteins are found in a cluster on chromosome 20. [provided by RefSeq
Sequence: MKSIILFVLSLLLILEKQAAVMGQKGGSKGQLPSGSSQFPHGKKGQHYFGQKDQQHTKSKGSFSIQHTYHVDINDHDWTRKSQQYDLNALHKATKSKQHLGGSQQLLNYKQEGRDHDKSKGHFHMIVIHHKGGQAHHGTQNPSQDQGNSPSGKGLSSQCSNTEKRLWVHGLSKEQASASGAQKGRTQGGSQSSYVLQTEELVVNKQQRETKNSHQNKGHYQNVVDVREEHSSKLQTSLHPAHQDRLQHGPKDIFTTQDELLVYNKNQHQTKNLNQDQEHGRKAHKISYPSSRTEERQLHHGEKSVQKDVSKGSISIQTEEKIHGKSQNQVTIHSQDQEHGHKENKISYQSSSTEERHLNCGEKGIQKGVSKGSISIQTEEQIHGKSQNQVRIPSQAQEYGHKENKISYQSSSTEERRLNSGEKDVQKGVSKGSISIQTEEKIHGKSQNQVTIPSQDQEHGHKENKMSYQSSSTEERRLNYGGKSTQKDVSQSSISFQIEKLVEGKSQIQTPNPNQDQWSGQNAKGKSGQSADSKQDLLSHEQKGRYKQESSESHNIVITEHEVAQDDHLTQQYNEDRNPISTSpecifications
| SEMG2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human SEMG2 protein. | |
| SEMG2 | |
| SGII | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6407 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC070306.1 | |
| SEMG2 | |
| SEMG2 (AAH70306.1,1 a.a. ∽ 582 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only