missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sema7a, Mouse, Clone: SKK-7, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of Sema7a.
Brand: Abnova MAB6582.25ug
This item is not returnable.
View return policy
Specifications
| Sema7a | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1.0-10 ug/mL) Flow Cyt (50 ng/mL) Immunoprecipitation (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| Sema7a | |
| Sema7a | |
| A synthetic peptide corresponding to extracellular domain of mouse Sema7a. | |
| RUO | |
| 20361 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Flow Cytometry, Immunoprecipitation | |
| SKK-7 | |
| Mouse monoclonal antibody raised against synthetic peptide of Sema7a. | |
| In PBS (0.1% proclin, 2% Block Ace) | |
| 2900057C09Rik/CDw108/H-Sema-L/M-Sema-L/Semal | |
| Mouse | |
| 25 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction