Learn More
SAT, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SAT.
Brand: Abnova H00006303-A01.50uL
Description
The protein encoded by this gene belongs to the acetyltransferase family, and is a rate-limiting enzyme in the catabolic pathway of polyamine metabolism. It catalyzes the acetylation of spermidine and spermine, and is involved in the regulation of the intracellular concentration of polyamines and their transport out of cells. Defects in this gene are associated with keratosis follicularis spinulosa decalvans (KFSD). Alternatively spliced transcripts have been found for this gene
Sequence: LLYLEDFFVMSDYRGFGIGSEILKNLSQVAMRCRCSSMHFLVAEWNEPSINFYKRRGASDLSSEEGWRLFKIDKEYLLKMATEESpecifications
| SAT | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SAT. | |
| SAT1 | |
| DC21/KFSD/SAT/SSAT/SSAT-1 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_002970 | |
| SAT1 | |
| SAT (NP_002961, 88 a.a. ∽ 171 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 6303 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only