Learn More
S100A11, Mouse, Clone: YK-1A4, Abnova™
Mouse monoclonal antibody raised against full-length recombinant S100A11.
Brand: Abnova MAB10144.100ug
Description
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in motility, invasion, and tubulin polymerization. Chromosomal rearrangements and altered expression of this gene have been implicated in tumor metastasis. [provided by RefSeq
Specifications
| S100A11 | |
| Monoclonal | |
| 1 mg/mL | |
| Mouse monoclonal antibody raised against full length recombinant S100A11. | |
| In 20mM PBS, pH 7.5, 300mM NaCl (50% glycerol) | |
| MLN70/S100C | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 6282 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Western Blot | |
| YK-1A4 | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| S100A11 | |
| S100A11 | |
| Recombinant protein corresponding to full length human S100A11. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.