missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RTN3, Mouse, Clone: 10A8, Abnova™
Mouse monoclonal antibody raised against recombinant RTN3.
Brand: Abnova MAB10363.100uL
This item is not returnable.
View return policy
Description
The reticulons are a group of highly conserved genes with preferential expression in neuroendocrine tissues (see, e.g., RTN1; MIM 600865).[supplied by OMIM
Specifications
| RTN3 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| RTN3 | |
| RTN3 | |
| Recombinant protein corresponding to human RTN3. | |
| RUO | |
| 10313 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a |
| ELISA, Immunohistochemistry | |
| 10A8 | |
| Mouse monoclonal antibody raised against recombinant RTN3. | |
| In ascites (0.03% sodium azide) | |
| ASYIP/HAP/NSPL2/NSPLII/RTN3-A1 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction