missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rotavirus p42, Mouse, Clone: 1-O-15, Abnova™
Mouse monoclonal antibody raised against rotavirus p42.
Brand: Abnova MAB5397.100ug
This item is not returnable.
View return policy
Specifications
| Rotavirus p42 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (5-10 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Virus | |
| Liquid |
| ELISA, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 1-O-15 | |
| Mouse monoclonal antibody raised against rotavirus p42. | |
| In PBS, pH 7.2 (0.01% sodium azide) | |
| Native purified bovine rotavirus virus. | |
| RUO | |
| Rotavirus p42 antigen. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction