Learn More
RNF4, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant RNF4.
Brand: Abnova H00006047-A01.50uL
Description
The protein encoded by this gene contains a RING finger motif and acts as a transcription regulator. This protein has been shown to interact with, and inhibit the activity of, TRPS1, a transcription suppressor of GATA-mediated transcription. Transcription repressor ZNF278/PATZ is found to interact with this protein, and thus reduce the enhancement of androgen receptor-dependent transcription mediated by this protein. Studies of the mouse and rat counterparts suggested a role of this protein in spermatogenesis. [provided by RefSeq
Sequence: YVTTHTPRNARDEGATGLRPSGTVSCPICMDGYSEIVQNGRLIVSTECGHVFCSQCLRDSLKNANTCPTCRKKINHKRYHPIYISpecifications
| RNF4 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant RNF4. | |
| RNF4 | |
| RES4-26/SNURF | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_002938 | |
| RNF4 | |
| RNF4 (NP_002929, 107 a.a. ∽ 190 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 6047 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only