Learn More
RNF167, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant RNF167.
Brand: Abnova H00026001-A01.50uL
Description
RNF167 is an E3 ubiquitin ligase that interacts with TSSC5 (SLC22A18; MIM 602631) and, together with UBCH6 (UBE2E1; MIM 602916), facilitates TSSC5 polyubiquitylation (Yamada and Gorbsky, 2006 [PubMed 16314844]).[supplied by OMIM
Sequence: VNGSVFIALLRRFDCNFDLKVLNAQKAGYGAAVVHNVNSNELLNMVWNSEEIQQQIWIPSVFIGERSSEYLRALFVYEKGARVLLVPDNTFPLGYSpecifications
| RNF167 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant RNF167. | |
| RNF167 | |
| 5730408C10Rik/DKFZp566H073/LP2254/RING105 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_015528 | |
| RNF167 | |
| RNF167 (NP_056343, 78 a.a. ∽ 172 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 26001 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only