Learn More
RIOK3, Mouse, Clone: 3G11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant RIOK3.
Brand: Abnova H00008780-M02A.200uL
Description
This gene was identified by the similarity of its product to the Aspergillus nidulans SUDD protein, an extragenic suppressor of the heat-sensitive bimD6 mutation that fails to attach properly to the spindle microtubules at a restrictive temperature. The specific function of this gene has not yet been determined. [provided by RefSeq
Sequence: NMLWHAGKVWLIDVSQSVEPTHPHGLEFLFRDCRNVSQKGGVKEALSERELFNAVSGLNITADNEADFLAEIEALEKMNEDHVQKNGRKAASFLKDDGDPPLLYDESpecifications
| RIOK3 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC039729 | |
| RIOK3 | |
| RIOK3 (AAH39729,411 a.a. ∽ 516 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 8780 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 3G11 | |
| Mouse monoclonal antibody raised against a partial recombinant RIOK3. | |
| RIOK3 | |
| DKFZp779L1370/SUDD | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.