missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHO, Mouse, Clone: 1D4, Abnova™
Mouse monoclonal antibody raised against full-length native RHO.
Brand: Abnova MAB8559.100ug
This item is not returnable.
View return policy
Specifications
| RHO | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:1000) Immunoprecipitation(1-2 ug/100-500 ug protein) The optimal working dilution should be determined by the end user. | |
| RHO | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody is specific to RHO. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 1D4 | |
| Mouse monoclonal antibody raised against full length native RHO. | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| RHO | |
| Native purified bovine RHO. | |
| RUO | |
| 509933 | |
| Bovine, Mammalia | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction