Learn More
REN, Mouse, Clone: 243/3 JG4, Abnova™
Mouse monoclonal antibody raised against REN.
Brand: Abnova MAB3697.1mg
Description
Renin catalyzes the first step in the activation pathway of angiotensinogen--a cascade that can result in aldosterone release,vasoconstriction, and increase in blood pressure. Renin, an aspartyl protease, cleaves angiotensinogen to form angiotensin I, which is converted to angiotensin II by angiotensin I converting enzyme, an important regulator of blood pressure and electrolyte balance. Transcript variants that encode different protein isoforms and that arise from alternative splicing and the use of alternative promoters have been described, but their full-length nature has not been determined. Mutations in this gene have been shown to cause familial hyperproreninemia. [provided by RefSeq
Specifications
| REN | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| REN | |
| REN | |
| Human REN. | |
| RUO | |
| 5972 | |
| Human | |
| Liquid |
| ELISA | |
| 243/3 JG4 | |
| Mouse monoclonal antibody raised against REN. | |
| In BBS | |
| FLJ10761 | |
| Mouse | |
| 1 mg | |
| Primary | |
| This antibody recognizes recombinant human renin. | |
| Store at 4°C. | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.