Learn More
REG3A, Mouse, Clone: 1H4, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant REG3A.
Brand: Abnova H00005068-M07.100ug
Description
This gene encodes a pancreatic secretory protein that may be involved in cell proliferation or differentiation. It has similarity to the C-type lectin superfamily. The enhanced expression of this gene is observed during pancreatic inflammation and liver carcinogenesis. Multiple alternatively spliced transcript variants encoding the same protein have been described for this gene but the full length nature of some transcripts is not yet known. [provided by RefSeq
Sequence: MLPPMALPSVSWMLLSCLMLLSQVQGEEPQRELPSARIRCPKGSKAYGSHCYALFLSPKSWTDADLACQKRPSGNLVSVLSGAEGSFVSSLVKSIGNSYSYVWIGLHDPTQGTEPNGEGWEWSSSDVMNYFAWERNPSTISSPGHCASLSRSTAFLRWKDYNCNVRLPYVCKFTDSpecifications
| REG3A | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC036776.1 | |
| REG3A | |
| REG3A (AAH36776.1,1 a.a. ∽ 175 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA | |
| 1H4 | |
| Mouse monoclonal antibody raised against a full-length recombinant REG3A. | |
| REG3A | |
| HIP/INGAP/PAP/PAP-H/PAP1/PBCGF/REG-III/REG3 | |
| Mouse | |
| Protein A Purification | |
| RUO | |
| 5068 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.