missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Tumors, epithelial origin, Mouse, Clone: CC52, Abnova™
Mouse monoclonal antibody raised against Rat Tumors, epithelial origin.
Brand: Abnova MAB5199.100ug
This item is not returnable.
View return policy
Specifications
| Rat Tumors, epithelial origin | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Rat | |
| Liquid |
| Western Blot | |
| CC52 | |
| Mouse monoclonal antibody raised against Rat Tumors, epithelial origin. | |
| In buffer containing 0.05% sodium azide | |
| Native purified from rat colon adenocarcinoma cells (CC531). | |
| RUO | |
| CC52 is a rat strain-independent marker for tumor cells of epithelial origin, such as colon, breast, or lung cancer, etc. When injected in colon tumor-bearing rats, CC52 localized to tumor cells (Hagenaars et al., 2001). CC52 can be used for immunohistochemical staining of tumor cells in frozen tissue sections as well as formalin-fixed/paraffin-embedded tissue. CC52 is also suitable for flow cytometry, Western blotting, immunoprecipitation, and ELISA. Hybridomas were selected for producing antibodies reacting with the CC531 cell line and with minimal cross reactivity to normal tissues (Hagenaars et al., 2001). CC52 belongs to a first generation of such antibodies and is suitable to detect tumor cells by immunohistochemistry. The monoclonal antibody binds to a dimer of two proteins, 120 kD, 130 kD, expressed by rat tumor cells of epithelial origin. CC52 has been used in many studies in a rat model of colorectal cancer (Hagenaars et al., 2000). In immunohistochemical stainings, there is | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction