missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rat Kappa Light Chain Secondary Antibody, FITC, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen A15426
Code : NEW
Additional Details : Weight : 0.00280kg
Description
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Rat Kappa Light Chain | |
Monoclonal | |
0.5 mg/mL | |
PBS with <0.1% sodium azide | |
Mouse | |
purified | |
RUO | |
Rat | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry | |
K4F5 | |
FITC | |
Ig kappa chain, Igk, Ig-kappa | |
This mouse IgG1-kappa class-switch variant monoclonal antibody reacts with rat kappa light chains and can be used for identification and enumeration of rat kappa light chain+ cells by flow cytometry. | |
100 μg | |
Secondary | |
4° C, store in dark | |
IgG2a κ |