Learn More
Mouse anti-Rat IgM, Clone: G53-238, Secondary Antibody, BD
Brand: BD Biosciences 553885
Additional Details : Weight : 0.01000kg
Description
The G53-238 antibody reacts specifically with rat IgM monomers and pentamers. It does not react with other Ig isotypes. G53-238 antibody has not been shown to stimulate B-cell proliferation.
Host Species: Mouse
Clone: G53-238
Isotype: IgG1, κ
Species Reactivity: Rat
Immunogen: Pooled rat Ig
ELISA, Flow Cytometry
Specifications
Mouse anti-Rat IgM | |
G53-238 | |
Mouse | |
0.5 mg | |
Immunology | |
Rat |
Monoclonal | |
0.5mg/mL | |
Affinity Purified | |
RUO | |
Secondary | |
IgG1 κ |