missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse anti-Rat IgG2b Secondary Antibody, Biotin
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
| Antigen | Rat IgG2b |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | MARG2b-8 |
| Concentration | 1 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide |
| Host Species | Mouse |
| Immunogen | IR863 rat IgG2b myeloma protein. |
| Purification Method | Affinity chromatography |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?