Learn More
Mouse anti-Rat IgG2b (Heavy chain) Secondary Antibody, FITC, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen SA125267
203.30 GBP valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Code : LD
Additional Details : Weight : 0.01000kg
Description
SA1-25267 detects rat IgG2b, Heavy Chain in flow cytometry applications. Store antibody at 4oC short term, and -20oC long term. Antibody is photosensitive; store in the dark.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Rat IgG2b (Heavy chain) | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide | |
Rat IgG2b, Heavy Chain | |
125 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG1 |
Flow Cytometry | |
MARG2b-3 | |
FITC | |
Mouse | |
purified | |
RUO | |
Rat | |
Liquid |