Learn More
Mouse anti-Rat IgG2b, Clone: RG7/11.1, Secondary Antibody, BD
Brand: BD Biosciences 553897
Additional Details : Weight : 0.01000kg
Description
The RG7/11.1 antibody reacts specifically with the Fc region of rat IgG2b. It does not react with other Ig isotypes.
Host Species: Mouse
Clone: RG7/11.1
Isotype: IgG2b, κ
Species Reactivity: Rat
Immunogen: Pooled rat IgG
ELISA, Flow Cytometry
Specifications
Mouse anti-Rat IgG2b | |
RG7/11.1 | |
Mouse | |
0.5 mg | |
Immunology | |
Rat |
Monoclonal | |
0.5mg/mL | |
Affinity Purified | |
RUO | |
Secondary | |
IgG2b κ |