missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse anti-Rat IgG1 Secondary Antibody, Biotin
Description
SA1-25262 recognizes rat IgG1. No reactivity with rat IgG2a, IgG2b, IgG2c, IgA, IgM, IgE, or IgD.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
| Antigen | Rat IgG1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | MARG1-2 |
| Concentration | 1 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide |
| Host Species | Mouse |
| Immunogen | Rat IR27 IgG1 myeloma protein. |
| Purification Method | purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?